missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC4C Monoclonal Antibody (1G5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522402
This item is not returnable.
View return policy
Description
Peptide Sequence: AVMLVIFYKM RKQHHRQNHH APTRTVEIIN VDDEITGDTP MESHLPMPAI EHEHLNHYNS YKSPFNHTTT VNTINSIHSS VHEPLLIRMN SKDNVQETQI.
Leucine-rich repeat-containing 4C (LRRC4C), also known as Netrin-G ligand 2 (NGL-2), is a gene highlighted for its expression in the nervous system, particularly the brain. It is a member of the leucine-rich repeat superfamily, which plays a significant role in cell adhesion and signaling pathways critical for synaptic connectivity and neuronal development. LRRC4C is involved in neural processes such as axon guidance and synapse formation, aiding in the establishment of functional neural circuits. Additionally, research has suggested that LRRC4C may function as a tumor suppressor, particularly in gliomas, indicating its potential role in neuro-oncological disorders. Variations or deletions in this gene have been linked to neurodevelopmental disorders, demonstrating its importance in normal brain function and development. Furthermore, LRRC4C's expression patterns and associations with cancer also suggest its potential as a prognostic marker in clinical settings.
Specifications
| LRRC4C | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9HCJ2 | |
| LRRC4C | |
| LRRC4C (NP_065980.1, 541 a.a. ~ 640 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1G5 | |
| Unconjugated | |
| LRRC4C | |
| 6430556C10Rik; KIAA1580; leucine rich repeat containing 4C; leucine-rich repeat-containing protein 4C; Lrrc4c; Netrin-G1 ligand; NGL1; NGL-1; RGD1311013; UNQ292/PRO331 | |
| Mouse | |
| Protein A | |
| RUO | |
| 57689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur