missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC47 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23217-1-AP-150UL
This item is not returnable.
View return policy
Description
LRRC47 gene ontology annotations related to this gene include phenylalanine-tRNA ligase activity; RNA binding.Specifications
| LRRC47 | |
| Polyclonal | |
| Unconjugated | |
| LRRC47 | |
| KIAA1185, LRRC47 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362672, 57470, 72946 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q505F5, Q8N1G4 | |
| Lrrc47 | |
| LRRC47 Fusion Protein Ag19736 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction