missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC43 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | LRRC43 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18247282
|
Novus Biologicals
NBP2-55428 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616156
|
Novus Biologicals
NBP2-55428-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
LRRC43 Polyclonal specifically detects LRRC43 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifikationer
| LRRC43 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| leucine rich repeat containing 43, leucine-rich repeat-containing protein 43, MGC35140 | |
| LRRC43 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 254050 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SELSVISGPSTILQMPRASAEELAKLRLRIDPRLCPSPGTVLFSTAHKPWAEVIPCSYEMQHSLRDLVPLKAFLLAGTTVTIVEEKILSWP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel