missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566692
This item is not returnable.
View return policy
Description
Immunogen sequence: GIGYLFSFRKL NCLDISGTGL KDIKTVKHKL QTHIGLVHSK VPLKEFDHSN CKTEGWADQI VLQWERVTAE AVKPR Highest antigen sequence identity to the following orthologs - mouse 88%, rat 91%.
LRRC42 is a protein coding gene.
Specifications
| LRRC42 | |
| Polyclonal | |
| Unconjugated | |
| LRRC42 | |
| A930011F22Rik; AA536996; dJ167A19.4; leucine rich repeat containing 42; leucine-rich repeat-containing protein 42; LRRC42; MGC137941 protein; RGD1308919 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y546 | |
| LRRC42 | |
| Recombinant Human LRRC42. Recombinant protein control fragment (Product #RP-107346). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction