missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562518
This item is not returnable.
View return policy
Description
Immunogen sequence: KPLKRFKRAA GKKGARTRQG PGAESEDLYD FVFIVAGEKE DGEEMEIGEV ACGALDGSDP SCLGLPALEA SQRF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Probable substrate recognition component of an ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. [UniProt].
Specifications
| LRRC41 | |
| Polyclonal | |
| Unconjugated | |
| LRRC41 | |
| AA409966; AW555107; D630045E04Rik; D730026A16Rik; elongin BC-interacting leucine-rich repeat protein; leucine rich repeat containing 41; leucine-rich repeat-containing protein 41; Lrrc41; Muf1; MUF1 protein; MUF1 protein (MUF1); PP7759; Protein Muf1; RGD1311221; RP4-636H5.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15345 | |
| LRRC41 | |
| Recombinant protein corresponding to Human LRRC41. Recombinant protein control fragment (Product #RP-99793). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction