missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LRRC3B/LRP15 Polyclonal antibody specifically detects LRRC3B/LRP15 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | LRRC3B/LRP15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | leucine rich repeat containing 3B, leucine-rich repeat-containing protein 3B, LRP15Leucine-rich repeat protein LRP15, MGC102927 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human LRRC3B/LRP15 (Accession#: NP_443185.1; Met1-Tyr204) |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?