missing translation for 'onlineSavingsMsg'
Learn More

LRRC39 Antibody (OTI2D9), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18427667 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427667 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427667 Supplier Novus Biologicals Supplier No. NBP272533AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRC39 Monoclonal antibody specifically detects LRRC39 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC39
Applications Western Blot
Classification Monoclonal
Clone OTI2D9
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Densin hlg, DKFZp313O1122, leucine rich repeat containing 39, leucine-rich repeat-containing protein 39, MGC14816
Host Species Mouse
Immunogen Full length recombinant protein of human LRRC39 (NP_653221) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127495
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.