missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32/GARP Antibody (855151) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6055JF669
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| LRRC32/GARP | |
| Monoclonal | |
| Janelia Fluor 669 | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| recombinant human LRRC32/GARP, Accession # Q14392 | |
| RUO | |
| 2615 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 855151 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction