missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA538058
This item is not returnable.
View return policy
Description
A recommended positive control is human prostate tissue lysate.
The LRRC26 protein was initially identified as a cytokeratin-associated protein whose expression was elevated in multiple cancer cell lines and cancer specimens as well in normal prostate and salivary gland tissues. LRRC26 was later found to negatively regulate the NF-kappaB activation and suppresses tumor growth and metastasis. LRRC26 can also act to selectively alter the efficacy of large conductance calcium-activated K (BK) channels.
Specifications
| LRRC26 | |
| Polyclonal | |
| Unconjugated | |
| LRRC26 | |
| bA350O14.10; BK channel auxiliary gamma subunit LRRC26; BK channel auxilliary gamma subunit LRRC26; CAPC; cytokeratin associated protein; cytokeratin-associated protein in cancer; leucine rich repeat containing 26; leucine-rich repeat-containing protein 26; Lrrc26; RGD1308398 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 389816 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q2I0M4 | |
| LRRC26 | |
| 18 amino acid peptide near the carboxy terminus of human LRRC26. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction