missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LRRC25 Monoclonal antibody specifically detects LRRC25 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
Specifications
| Antigen | LRRC25 |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI1A2 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | leucine rich repeat containing 25, leucine-rich repeat-containing protein 25, MAPAFLJ38116, monocyte and plasmacytoid activated molecule, Monocyte and plasmacytoid-activated protein |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human LRRC25 (NP_660299) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?