missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LRRC23 Overexpression Lysate

Product Code. 18281611 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18281611 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18281611 Supplier Novus Biologicals™ Supplier No. NBL112681
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The LRRC23 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC26514 and based on accession number NM_2165. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens leucine rich repeat containing 23 (LRRC23), transcript variant 1, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.