missing translation for 'onlineSavingsMsg'
Learn More

LRRC2 Antibody (1G3), Novus Biologicals™

Product Code. 18409948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409948 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409948 Supplier Novus Biologicals Supplier No. H00079442M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRC2 Monoclonal antibody specifically detects LRRC2 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1G3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_078788
Gene Alias leucine rich repeat containing 2, leucine-rich repeat-containing 2, leucine-rich repeat-containing protein 2
Host Species Mouse
Immunogen LRRC2 (NP_078788, 272 a.a. ∽ 371 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LTYLPYSMLNLKKLTLLVVSGDHLVELPTALCDSSTPLKFVSLMDNPIDNAQCEDGNEIMESERDRQHFDKEVMKAYIEDLKERESVPSYTTKVSFSLQL
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79442
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.