missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20918-1-AP-20UL
This item is not returnable.
View return policy
Description
Leucine-rich repeat-containing protein 17 (LRRC17) is an LRR protein predomitly expressed by osteoblasts. LRRC17 acts as a negative regulator of receptor activator of NF-kappaB ligand (RANKL)-induced osteoclast differentiation. It has been reported that low plasma level of LRRC17 is an independent risk factor for osteoporotic fractures.Specifications
| LRRC17 | |
| Polyclonal | |
| Unconjugated | |
| LRRC17 | |
| LRRC17, P37NB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10234, 74511 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N6Y2, Q9CXD9 | |
| Lrrc17 | |
| LRRC17 Fusion Protein Ag15117 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction