missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC16B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571874
This item is not returnable.
View return policy
Description
LRRC16B Polyclonal Antibody for Western Blot
CARMIL3 is a protein coding gene.
Specifications
| LRRC16B | |
| Polyclonal | |
| Unconjugated | |
| CARMIL3 | |
| C14orf121; capping protein regulator and myosin 1 linker 3; capping protein regulator and myosin 1 linker protein 3; Capping protein, Arp2/3 and myosin-I linker protein 3; CARMIL3; crml-1; leucine rich repeat containing 16B; leucine-rich repeat-containing protein 16B; LRRC16B; mFLJ00240 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 268747, 90668 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q3UFQ8, Q8ND23 | |
| CARMIL3 | |
| KLH conjugated synthetic peptide between 1344-1372 amino acids from the C-terminal region of human LRRC16B. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction