Learn More
Invitrogen™ LRRC15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113927
Description
Immunogen sequence: LNISALIALR IEKNELSRIT PGAFRNLGSL RYLSLANNKL QVLPIGLFQG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Leucine-rich repeats (LRRs) are 20-29 amino acid motifs that mediate protein-protein interactions. The primary function of these motifs is to provide a versatile structural framework for the formation of these protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervous system development. Several human diseases are associated with mutations in genes encoding LRR-containing proteins. The leucine-rich repeat-containing protein 15 (LRRC15, also designated LIB) is a 581 amino acid protein that contains 15 LRR repeats and is involved in cell-cell and/or -extracellular matrix interactions. LRRC15 is frequently overexpressed in multiple tumor types, most notably breast carcinoma. It is also associated with the pathogenesis of Alzheimer's disease.
Specifications
| LRRC15 | |
| Polyclonal | |
| Unconjugated | |
| LRRC15 | |
| 5430427N11Rik; hLib; leucine rich repeat containing 15; Leucine-rich repeat protein induced by amyloid-beta; leucine-rich repeat protein induced by beta amyloid; leucine-rich repeat protein induced by beta-amyloid; leucine-rich repeat protein induced by beta-amyloid homolog; Leucine-rich repeat-containing protein 15; LIB; Lrrc15; rLib | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 131578 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TF66 | |
| LRRC15 | |
| Synthetic peptide directed towards the N terminal region of human LRRC15 (aa 74-123). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.