missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570766
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Human: 100%; Rabbit: 91%.
LRRC10 may play important roles in cardiac development and/or cardiac function.
Specifications
| LRRC10 | |
| Polyclonal | |
| Unconjugated | |
| LRRC10 | |
| D330003I11Rik; Heart-restricted leucine-rich repeat protein; HRLRRP; leucine rich repeat containing 10; leucine-rich containing 10; leucine-rich repeat protein; leucine-rich repeat-containing 10; leucine-rich repeat-containing protein 10; LRRC10; LRRC10A; Serdin1; Unknown (protein for MGC:137420) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 376132 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5BKY1 | |
| LRRC10 | |
| synthetic peptide directed towards the C-terminal region of human LRRC10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction