missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10128-2-AP-150UL
This item is not returnable.
View return policy
Description
Lano is a member of the LAP (leucine-rich repeats and PDZ) family of proteins that also includes Densin-180, Erbin, and hScribble. The LAP proteins generally contain multiple leucine-rich repeat (LRR) domains which serve to target them to the basolateral membrane of epithelial cells. Lano is unique in that it alone does not possess one or more PDZ (PSD95/DLG/ZO-1) domains as do the other members of the LAP family. However, it can bind to the PDZ domain of Erbin in addition to those of membrane-associated and guanylate kinase (MAGUK) proteins which regulate adhesion and plasticity at cell junctions. It has been suggested that it is through these interaction that these LAP proteins participate in the maintece of proper embryonic development and integrity of epithelial tissues.Specifications
| LRRC1 | |
| Polyclonal | |
| Unconjugated | |
| LRRC1 | |
| LANO, LANO adapter protein, LAP and no PDZ protein, LRRC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 214345, 367113, 55227 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80VQ1, Q9BTT6 | |
| Lrrc1 | |
| LRRC1 Fusion Protein Ag0175 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction