missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRR1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567045

Product Code. 15909724

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LDDNELIQFPC KIGQLINLRF LSAARNKLPF LPSEFRNLSL EYLDLFGNTF EQPKVLPVIK LQAPLTLLES SART Highest antigen sequence identity to the following orthologs - mouse 76%, rat 72%.

May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRR1
Polyclonal
Unconjugated
LRR1
2410005L11Rik; 4-1BBlrr; 4-1BB-mediated signaling molecule; 4-1BB-mediated-signaling molecule; AU021780; cyclophilin-like 5; leucine rich repeat protein 1; leucine-rich repeat protein 1; LRR1; LRR-1; LRR-repeat protein 1; peptidylprolyl isomerase (cyclophilin) like 5; peptidylprolyl isomerase (cyclophilin)-like 5, isoform 3; peptidylprolyl isomerase-like 5; PPIL5
Rabbit
Antigen Affinity Chromatography
RUO
122769
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96L50
LRR1
Recombinant Human LRR1. Recombinant protein control fragment (Product #RP-105474).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.