missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRPAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24662-1-AP-20UL
This item is not returnable.
View return policy
Description
Interacts with LRP1/alpha-2-macroglobulin receptor and glycoprotein 330.Specifications
| LRPAP1 | |
| Polyclonal | |
| Unconjugated | |
| LRPAP1 | |
| A2MRAP, A2RAP, Alpha 2 MRAP, HBP44, LRPAP1, MRAP, RAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4043 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P30533 | |
| LRPAP1 | |
| LRPAP1 Fusion Protein Ag20304 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction