missing translation for 'onlineSavingsMsg'
Learn More

LRPAP Antibody (7F1), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18361783 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361783 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361783 Supplier Novus Biologicals Supplier No. NBP312046JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRPAP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 7F1
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP
Host Species Mouse
Immunogen This antibody was raised by immunising mice with human placental LRPAP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4043
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.