missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
Specifications
| Antigen | LRPAP |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 7F1 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP |
| Host Species | Mouse |
| Immunogen | This antibody was raised by immunising mice with human placental LRPAP. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?