missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109269
This item is not returnable.
View return policy
Description
Positive Control: Mouse placenat tissue, SH-SY-5Y, F9, HepG2, human placenta tissue.
Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
Specifications
| LRP8 | |
| Polyclonal | |
| Unconjugated | |
| LRP8 | |
| 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7804 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14114 | |
| LRP8 | |
| Recombinant protein within Human APOER2 aa 494-722. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction