Learn More
Invitrogen™ LRP8 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA587962
Product Code. 16596311
  • £441.00 / 100µL
Description
Immunogen sequence: PLWAEPCLGE TREPEDPAPA LKELFVLPGE PRSQLHQLPK NPLSELPVVK SKRVALSLED DGLP; Positive Samples: SH-SY5Y, Rat testis; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRP8
Polyclonal
Unconjugated
LRP8
4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B
Rabbit
Affinity Chromatography
RUO
362558, 7804
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.58 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14114
LRP8
A synthetic peptide corresponding to a sequence within amino acids 900 to the C-terminus of human ApoER2/ApoER2/LRP8 (NP_0046222).
100 μL
Primary
Human, Rat, Rat