missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116913
This item is not returnable.
View return policy
Description
PA1-16913 is specific for the ~106 kDa (membrane form) ApoER2 protein. A larger ~130 kDa band may also be seen (glycosylated form). Suggested positive control: mouse brain membrane preparation. Immunogen sequence homology: bovine (95%).
Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
Specifications
| LRP8 | |
| Polyclonal | |
| Unconjugated | |
| LRP8 | |
| 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16975, 396102, 7804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q14114, Q924X6, Q98931 | |
| LRP8 | |
| A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction