Learn More
Invitrogen™ LRP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589161
Description
Immunogen sequence: APLLLYANRR DLRLVDATNG KENATIVVGG LEDAAAVDFV FSHGLIYWSD VSEEAIKRTE FNKTESVQNV VVSGLLSPDG LACDWLGEKL YWTDSETNRI EVSNLDGSLR KVLFWQELDQ PRAIALDPSS G; Positive Samples: Mouse liver; Cellular Location: Endoplasmic reticulum, Membrane, Single-pass type I membrane protein.
LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.
Specifications
| LRP6 | |
| Polyclonal | |
| Unconjugated | |
| LRP6 | |
| ADCAD2; C030016K15Rik; Cd; crooked; FLJ90062; FLJ90421; Gw; LDL receptor related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; low-density lipoprotein receptor-related protein 6; lpr6; LRP6; LRP-6; ska>m26Jus>; ska26; skax26; STHAG7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16974, 4040 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75581, O88572 | |
| LRP6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-150 of human LRP6 (NP_0023272). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.