missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP5 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149231
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
LDL related proteins, LRP 5 and LRP 6, are members of the LDL receptor superfamily, which were found to be involved in the canonical Wnt signaling pathway. Mammalian LRP 6 was shown to bind to Wnt 1 and enhance Wnt-induced developmental processes in Xenopus embryos. Mice lacking LRP 6 exhibited developmental defects that are similar to those caused by deficiencies in various Wnt proteins. Recent work also shows that Wnt proteins induce the binding of LRP 5 to Axin, leading Axin degradation and beta catenin stabilization.
Specifications
| LRP5 | |
| Polyclonal | |
| Unconjugated | |
| LRP5 | |
| BMND1; EVR1; EVR4; HBM; HGNC:6697; LDL receptor related protein 5; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LR3; Lrp5; LRP-5; LRP7; LRP-7; mKIAA4142; OPPG; OPS; OPTA1; VBCH2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 4041 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O75197 | |
| LRP5 | |
| E. coli-derived recombinant human LRP-5, Phe1422-Ser1615. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction