missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118912
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LRP4.
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
Specifications
| LRP4 | |
| Polyclonal | |
| Unconjugated | |
| LRP4 | |
| 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 228357, 4038, 83469 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75096, Q8VI56, Q9QYP1 | |
| LRP4 | |
| A synthesized peptide derived from human LRP4(Accession O75096), corresponding to amino acid residues T271-N321. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction