missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP4 Polyclonal Antibody

LRP4 Polyclonal Antibody
Brand: Invitrogen PA547795
This item is not returnable.
View return policy
Description
LRP4 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| LRP4 | |
| Polyclonal | |
| Unconjugated | |
| LRP4 | |
| 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4038, 83469 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O75096, Q9QYP1 | |
| LRP4 | |
| E. coli-derived recombinant human LRP-4 Gly18-Ser344. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction