Learn More
Invitrogen™ LRP4 Monoclonal Antibody (N207/27)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527675
Description
1 μg/mL of MA5-27675 was sufficient for detection of LRP4 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects the extracellular domain of LRP4 approximately 215kDa. Degradation product can be seen at approximately 150kDa. This antibody was formerly sold as clone S207-27.
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
Specifications
| LRP4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| O75096, Q8VI56, Q9QYP1 | |
| LRP4 | |
| Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| N207/27 | |
| Unconjugated | |
| LRP4 | |
| 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 228357, 4038, 83469 | |
| -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.