missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LRP3 Antibody (7B3A5E9), Novus Biologicals™

Mouse Monoclonal Antibody

£217.00 - £389.00

Specifications

Antigen LRP3
Clone 7B3A5E9
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500
Applications Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30031441
View Documents
Novus Biologicals™
NBP3-27129-0.025ml
0.025 mL
£217.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30029786
View Documents
Novus Biologicals™
NBP3-27129-0.1ml
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LRP3 Monoclonal antibody specifically detects LRP3 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

LRP3
1 mg/mL
Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Mouse
Human
hLRp105, low density lipoprotein receptor-related protein 3,105 kDa low-density lipoprotein receptor-related protein, low-density lipoprotein receptor-related protein 3, LRP-3
Purified recombinant fragment of human LRP3 (AA: extra 43-184) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
7B3A5E9
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500
Monoclonal
Purified
RUO
PBS
4037
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.