missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRP130 Recombinant Rabbit Monoclonal Antibody (2V9I0)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557957

 View more versions of this product

Product Code. 30283096

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LWYEDEKHSL NSSSASTTEP DFQKDILIAC RLNQKKGAYD IFLNAKEQNI VFNAETYSNL IKLLMSEDYF TQAMEVKAFA ETHIKGFTLN DAANSRLIIT QVRRDYLKEA VTTLKTVLDQ QQTPSRLAVT RVIQALAMKG DVENIEVVQK MLNGLEDSIG LSKMVFINNI ALAQIKNNNI DAAIENIENM LTSENKVIEP QYFGLAYLFR KVIEEQLEPA VEKISIMAER LANQFAIYKP VTDFFLQLVD AGKVDDARAL LQRCGAIAEQ TPILLLFLLR NSRKQGKAST VKSVLELIPE LNEKEEAYNS LMKSYVSEKD VTSAKALYEH LTAKNTKLDD LFLKRYASLL KYAGEPVPFI EPPESFEFYA QQLRKLRENS S.

This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3' to 5' exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRP130
Recombinant Monoclonal
0.58 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P42704
LRPPRC
Recombinant fusion protein containing a sequence corresponding to amino acids 1014-1394 of human LRPPRC (NP_573566.2).
20 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
2V9I0
Unconjugated
LRPPRC
130 kDa leucine-rich protein; 3110001K13Rik; C76645; CLONE-23970; GP130; leucine rich pentatricopeptide repeat containing; leucine rich protein 157; leucine rich protein LRP130; leucine-rich pentatricopeptide repeat containing; leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; leucine-rich protein 157; LRP 130; LRP130; Lrp157; Lrpprc; LSFC; mitochondrial leucine-rich PPR motif-containing protein; rLRP157
Rabbit
Protein A
RUO
10128
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.