missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP130 Recombinant Rabbit Monoclonal Antibody (1H19L11)
LRP130 Recombinant Rabbit Monoclonal Antibody (1H19L11)
Brand: Invitrogen 703100
This item is not returnable.
View return policy
Description
LRP130 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, RIP
Specifications
| LRP130 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P42704 | |
| LRPPRC | |
| Protein corresponding to human LRP130 [aa611-aa907]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, RNA Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 1H19L11 | |
| Unconjugated | |
| LRPPRC | |
| 130 kDa leucine-rich protein; 3110001K13Rik; C76645; CLONE-23970; GP130; leucine rich pentatricopeptide repeat containing; leucine rich protein 157; leucine rich protein LRP130; leucine-rich pentatricopeptide repeat containing; leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; leucine-rich protein 157; LRP 130; LRP130; Lrp157; Lrpprc; LSFC; mitochondrial leucine-rich PPR motif-containing protein; rLRP157 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction