missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557643
This item is not returnable.
View return policy
Description
Immunogen sequence: RIWDTLQKLG AVYDVSHYNA LLKVYLQNEY KFSPTDFLAK MEEANIQPNR VTYQRLIASY CNVGDIEGAS KILGFMKTKD LPVT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.
This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3' to 5' exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.
Specifications
| LRP130 | |
| Polyclonal | |
| Unconjugated | |
| LRPPRC | |
| 130 kDa leucine-rich protein; 3110001K13Rik; C76645; CLONE-23970; GP130; leucine rich pentatricopeptide repeat containing; leucine rich protein 157; leucine rich protein LRP130; leucine-rich pentatricopeptide repeat containing; leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; leucine-rich protein 157; LRP 130; LRP130; Lrp157; Lrpprc; LSFC; mitochondrial leucine-rich PPR motif-containing protein; rLRP157 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42704 | |
| LRPPRC | |
| Recombinant protein corresponding to Human LRP130. Recombinant protein control fragment (Product #RP-96095). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction