missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP12 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148603
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Members of the LDL receptor gene family, including LDLR (low density lipoprotein receptor), LRP1 (low density lipoprotein related protein), Megalin (also designated GP330), VLDLR (very low density lipoprotein receptor) and ApoER2 are characterized by a cluster of cysteine-rich class A repeats, epidermal growth factor (EGF)-like repeats, YWTD repeats and an O-linked sugar domain. LRP12, also designated ST7, is a member of the LDLR family that is thought to be involved in the internalization of lipophilic molecules and/or signal transduction. LRP12 has been shown to interact with RACK1, MADHIP and Integrin beta-1-binding protein 3. It is widely expressed in heart, skeletal muscle, brain, lung, placenta and pancreas. Overexpression of LRP12 may be associated with oral tumors, therefore implicating LRP12 as a potential oncogene.
Specifications
| LRP12 | |
| Polyclonal | |
| Unconjugated | |
| LRP12 | |
| AI848829; C820005L12Rik; LDL receptor related protein 12; LDLR-related protein 12; low density lipoprotein receptor-related protein 12; low density lipoprotein-related protein 12; low-density lipoprotein receptor-related protein 12; LRP12; LRP-12; ST7; Suppressor of tumorigenicity 7 protein | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 29967 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y561 | |
| LRP12 | |
| Mouse myeloma cell line NS0-derived recombinant human ST7/LRP12, Asn28-Ile488. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction