missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRP11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16357864
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16357864 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16357864 Supplier Invitrogen™ Supplier No. PA580776

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LRP11.

LRP11 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP11
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LRP11
Gene Accession No. Q86VZ4
Gene Alias bA350J20.3; LDL receptor related protein 11; low density lipoprotein receptor-related protein 11; low-density lipoprotein receptor-related protein 11; LRP11; LRP-11; MANSC3
Gene Symbols LRP11
Host Species Rabbit
Immunogen Recombinant Human LRP11 Protein (Met1-Gly450).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84918
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.