missing translation for 'onlineSavingsMsg'
Learn More

LRP10 Antibody, Novus Biologicals™

Product Code. 18420261 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420261 0.1 mL 0.1mL
18497790 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18420261 Supplier Novus Biologicals Supplier No. NBP184159

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRP10 Polyclonal antibody specifically detects LRP10 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP10
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias DKFZp564C1940, low density lipoprotein receptor-related protein 10, low-density lipoprotein receptor-related protein 10, LRP-10, LRP9, MGC142274, MGC142276, MGC8675, MST087
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: PNHACEDPPAVLLEVQGTLQRPLVRDSRTSPANCTWLILGSKEQTVTIRFQKLHLACGSERLTLRSPLQPLISLCEAPPSPLQLPGGNVTITYSYAGARAPMGQGFLLSYSQDWLMCLQEEFQCLNHRCVSAVQRCDG
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 26020
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.