missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-6 Antibody (SR1271), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | LRP-6 |
|---|---|
| Clone | SR1271 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605964
|
Novus Biologicals
NBP3-22014-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602424
|
Novus Biologicals
NBP3-22014-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRP-6 Monoclonal antibody specifically detects LRP-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| LRP-6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
| A synthesized peptide derived from human LRP-6 (Uniprot #: O75581) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1271 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Stem Cell Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4040 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title