missing translation for 'onlineSavingsMsg'
Learn More

LRP-6 Antibody (SR1271), R&D Systems™

Product Code. 18602424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602424 50 μg 50µL
18605964 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18602424 Supplier Novus Biologicals Supplier No. NBP32201450ul
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

LRP-6 Monoclonal specifically detects LRP-6 in Human∣Mouse∣Rat samples. It is validated for Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-6
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SR1271
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6
Host Species Rabbit
Immunogen A synthesized peptide derived from human LRP-6 (Uniprot #: O75581)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Phospho Specific, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4040
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.