missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-6 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-1475
This item is not returnable.
View return policy
Description
LRP-6 Polyclonal specifically detects LRP-6 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| LRP-6 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize an epitope corresponding to aa 1546-1560 of human LRP6 protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry 10 ug/mL, Immunohistochemistry 4 - 6 ug/mL, Immunohistochemistry-Paraffin 4 - 6 ug/mL, Peptide ELISA Detection limit 1:64000 | |
| O75581 | |
| LRP6 | |
| Peptide with sequence CMTSVATAKGYTSDL corresponding to internal region according to NP_002327.2. | |
| 0.1 mg | |
| Stem Cell Signaling Pathway | |
| 4040 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction