missing translation for 'onlineSavingsMsg'
Learn More

LRP-5 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30735106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735106 Supplier Novus Biologicals Supplier No. AF6646L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

LRP-5 Polyclonal antibody specifically detects LRP-5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-5
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias BMND1OPTA1, EVR1, EVR4, HBM, low density lipoprotein receptor-related protein 5, low density lipoprotein receptor-related protein 7, low-density lipoprotein receptor-related protein 5, LR3VBCH2, LRP-5, LRP7exudative vitreoretinopathy 1, OPPG, OPS, osteoporosis pseudoglioma syndrome
Host Species Sheep
Immunogen E. coli-derived recombinant human LRP-5, Phe1422-Ser1615, Accession # O75197
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Stem Cell Signaling Pathway, Vision, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4041
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.