missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-4 Antibody (741704), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5948CL1
This item is not returnable.
View return policy
Description
LRP-4 Monoclonal antibody specifically detects LRP-4 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| LRP-4 | |
| Monoclonal | |
| CoraFluor 1 | |
| CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
| E. coli-derived recombinant human LRP-4, Gly18-Ser344, Accession # O75096 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 4038 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| 741704 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction