missing translation for 'onlineSavingsMsg'
Learn More

LRP-1B Antibody (1034124) [FITC], Novus Biologicals™

Product Code. 30735386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735386 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735386 Supplier Novus Biologicals Supplier No. FAB2368F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRP-1B Monoclonal antibody specifically detects LRP-1B in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-1B
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1034124
Conjugate FITC
Formulation PBS
Gene Alias EC 1.1.1.94, EC 3.4.21.9, low density lipoprotein receptor related protein-deleted in tumor, low density lipoprotein receptor-related protein 1B, low density lipoprotein-related protein 1B (deleted in tumors), Low-density lipoprotein receptor-related protein-deleted in tumor, LRP-1B, LRP-deleted in tumors, LRPDIT, LRP-DITlow-density lipoprotein receptor-related protein 1B
Host Species Mouse
Immunogen E. coli-derived human LRP-1 Cluster II, Gln4449-Ala4544, Accession # Q07954
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 53353
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.