missing translation for 'onlineSavingsMsg'
Learn More

LRP-1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18407766 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407766 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407766 Supplier Novus Biologicals Supplier No. NBP262753AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRP-1 Polyclonal antibody specifically detects LRP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias A2MR, Alpha-2-Macroglobulin Receptor, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, LRP1A, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, TGFBR5, type V tgf-beta receptor
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal of Human Low-density lipoprotein receptor-related protein 1 (aa. 160-260)
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4035
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.