missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRMP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LRMP |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859024
|
Proteintech
19498-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849024
|
Proteintech
19498-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays a role in the delivery of peptides to major histocompatibility complex (MHC) class I molecules; this occurs in a transporter associated with antigen processing (TAP)-independent manner. May play a role in taste signal transduction via ITPR3. May play a role during fertilization in pronucleus congression and fusion.Specifications
| LRMP | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q12912 | |
| 4033 | |
| Peptide | |
| Primary | |
| -20°C | |
| LRMP |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| JAW1, LRMP, Protein Jaw1 | |
| LRMP | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title