missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lrmp Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549246
This item is not returnable.
View return policy
Description
Lrmp Polyclonal Antibody for Western Blot
Plays a role in the delivery of peptides to major histocompatibility complex (MHC) class I molecules; this occurs in a transporter associated with antigen processing (TAP)-independent manner. May play a role in taste signal transduction via ITPR3. May play a role during fertilization in pronucleus congression and fusion.
Specifications
| Lrmp | |
| Polyclonal | |
| Unconjugated | |
| LRMP | |
| D6Int3; D6Int4; D6Int5; D6Int7; D6Int8; JAW1; LRMP; lymphoid restricted membrane protein; lymphoid-restricted membrane protein; Processed lymphoid-restricted membrane protein; Protein Jaw1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 16970 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q60664 | |
| LRMP | |
| KLH conjugated synthetic peptide between 403-431 amino acids from the C-terminal region of mouse Lrmp. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction