Learn More
Invitrogen™ Lrmp Monoclonal Antibody (K52026_19B2), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | Lrmp |
|---|---|
| Clone | K52026_19B2 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30211026
|
Invitrogen™
MA555759 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204577
|
Invitrogen™
MA556139 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: LSGCIRMNDD PSMEENGVER VCPESLLQSR EYSSLPLPRH TSSTDGTITS SDPGLEILNM ASCDLDRNSL CKKEEDTRSA SPTIEAQGTS PAHDNIAFQD STSKDKTILN LEAKEEPETI EEHKKEHASG DSVVSPLPVT TVKSVNLRQS ENTSANEKEV EAEFLRLSLG FKCDWFTLEK RVKLEERSRD LAEENLKKEI TNCLKLLESL TPLCEDDNQA QEIIKKLEKS IKFLSQCAAR VASRAEMLGA INQESRVSKA VEVMIQHVEN LKRMYAKEHA ELEELKQVLL QNERSFNPLE
Lrmp Monoclonal antibody specifically detects Lrmp in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| Lrmp | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| D6Int3; D6Int4; D6Int5; D6Int7; D6Int8; JAW1; LRMP; lymphoid restricted membrane protein; lymphoid-restricted membrane protein; Processed lymphoid-restricted membrane protein; Protein Jaw1 | |
| Recombinant human IRAG2 (immunogen range is 51-350). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LRMP |
| K52026_19B2 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q12912 | |
| 4033 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.