missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lrmp Monoclonal Antibody (K52026_16E2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556348

 View more versions of this product

Product Code. 30199694

  • £760.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: LSGCIRMNDD PSMEENGVER VCPESLLQSR EYSSLPLPRH TSSTDGTITS SDPGLEILNM ASCDLDRNSL CKKEEDTRSA SPTIEAQGTS PAHDNIAFQD STSKDKTILN LEAKEEPETI EEHKKEHASG DSVVSPLPVT TVKSVNLRQS ENTSANEKEV EAEFLRLSLG FKCDWFTLEK RVKLEERSRD LAEENLKKEI TNCLKLLESL TPLCEDDNQA QEIIKKLEKS IKFLSQCAAR VASRAEMLGA INQESRVSKA VEVMIQHVEN LKRMYAKEHA ELEELKQVLL QNERSFNPLE

Plays a role in the delivery of peptides to major histocompatibility complex (MHC) class I molecules; this occurs in a transporter associated with antigen processing (TAP)-independent manner. May play a role in taste signal transduction via ITPR3. May play a role during fertilization in pronucleus congression and fusion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lrmp
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q12912
LRMP
Recombinant human IRAG2 (immunogen range is 51-350).
200 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K52026_16E2
Unconjugated
LRMP
D6Int3; D6Int4; D6Int5; D6Int7; D6Int8; JAW1; LRMP; lymphoid restricted membrane protein; lymphoid-restricted membrane protein; Processed lymphoid-restricted membrane protein; Protein Jaw1
Mouse
Protein A
RUO
4033
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.