missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRMP/Jaw1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | LRMP/Jaw1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18360916
|
Novus Biologicals
NBP3-03425-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18386116
|
Novus Biologicals
NBP3-03425-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRMP/Jaw1 Polyclonal specifically detects LRMP/Jaw1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| LRMP/Jaw1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 4033 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| JAW1Protein Jaw1, lymphoid-restricted membrane protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human LRMP/Jaw1 (NP_006143.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title