missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRIG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534865
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Chicken (87%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in craniofacial and inner ear morphogenesis during embryonic development. May act within the otic vesicle epithelium to control formation of the lateral semicircular canal in the inner ear, possibly by restricting the expression of NTN1.
Specifications
| LRIG3 | |
| Polyclonal | |
| Unconjugated | |
| LRIG3 | |
| leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6UXM1 | |
| LRIG3 | |
| Recombinant protein encompassing a sequence within the center region of human LRIG3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction