missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRIG1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18504CL1
This item is not returnable.
View return policy
Description
LRIG1 Polyclonal antibody specifically detects LRIG1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| LRIG1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| D6Bwg0781e, Img, LIG-1 | |
| Recognizes region between residues Between between 1043 and 1093.. | |
| 0.1 mL | |
| Cancer | |
| 26018 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction